Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260763.1 | NP_001247692.1 | 913 | 139 | ||
XM_015140181.2 | XP_014995667.1 | 1242 | 139 | |||
XM_015140182.2 | XP_014995668.1 | 1003 | 139 | |||
XM_015140184.2 | XP_014995670.1 | 1280 | 139 | |||
XM_028849138.1 | XP_028704971.1 | 1030 | 139 | |||
XM_028849139.1 | XP_028704972.1 | 1109 | 139 | |||
XM_028849140.1 | XP_028704973.1 | 1575 | 139 | |||
GenBank mRNA | JU336108.1 | - | 1001 | 139 | ||
JU476968.1 | - | 1262 | 139 | |||
JU476969.1 | - | 982 | 139 | |||
JV048063.1 | - | 913 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
