Assay Details
| Gene Symbol: GET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132874.2 | XP_014988360.1 | 566 | 103 | ||
| GenBank mRNA | JU319627.1 | - | 545 | 103 | ||
| JU477093.1 | - | 521 | 103 | |||
| JV045214.1 | - | 482 | 103 | |||
| JV695474.1 | - | 519 | 103 | |||
More Information
Set Membership: |
|

GET1
Genomic Map