Assay Details
| Gene Symbol: PCDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141091.2 | XP_014996577.1 | 3452 | 63 | ||
| XM_028849808.1 | XP_028705641.1 | 3927 | 63 | |||
| GenBank mRNA | JU327485.1 | - | 3298 | 63 | ||
| JV583375.1 | - | 3248 | 63 | |||
More Information
Set Membership: |
|

PCDH1
Genomic Map