Assay Details
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110224.2 | XP_014965710.1 | 3446 | 125 | ||
XM_028830749.1 | XP_028686582.1 | 1203 | 125 | |||
GenBank mRNA | JU497237.1 | - | 223 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
