Assay Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086657.3 | XP_001086657.1 | 2579 | 58 | ||
| XM_015143720.2 | XP_014999206.1 | 2653 | 58 | |||
| XM_015143721.2 | XP_014999207.1 | 2177 | 58 | |||
| XM_015143723.2 | XP_014999209.1 | 972 | 58 | |||
| XM_028851560.1 | XP_028707393.1 | 2601 | 58 | |||
| XM_028851561.1 | XP_028707394.1 | 2315 | 58 | |||
| XM_028851562.1 | XP_028707395.1 | 892 | 58 | |||
| GenBank mRNA | JU321323.1 | - | 1781 | 58 | ||
| JU321475.1 | - | 2045 | 58 | |||
More Information
Set Membership: |
|

SAMD4A
Genomic Map