Assay Details
Gene Symbol: INKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148287.2 | XP_015003773.1 | 236 | 79 | ||
GenBank mRNA | JU322528.1 | - | 195 | 79 | ||
JU473219.1 | - | 189 | 79 | |||
JV600664.1 | - | 193 | 79 | |||
JV668568.1 | - | 238 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
