Assay Details
Gene Symbol: SMIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099113.4 | XP_001099113.1 | 300 | 61 | ||
XM_015120390.2 | XP_014975876.1 | 1008 | 61 | |||
GenBank mRNA | JU322817.1 | - | 576 | 61 | ||
JU322818.1 | - | 571 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
