Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134238.2 | XP_014989724.1 | 636 | 92 | ||
| XM_015134239.2 | XP_014989725.1 | 713 | 92 | |||
| GenBank mRNA | JU336652.1 | - | 461 | 92 | ||
| JU494653.1 | - | 442 | 92 | |||
More Information
Set Membership: |
|

RBM48
Genomic Map