Assay Details
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131882.2 | XP_014987368.1 | 612 | 68 | ||
XM_015131883.2 | XP_014987369.1 | 612 | 68 | |||
GenBank mRNA | JU490891.1 | - | 117 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
