Assay Details
Gene Symbol: PABPC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848770.1 | XP_028704603.1 | 1248 | 96 | ||
XM_028848771.1 | XP_028704604.1 | 1193 | 96 | |||
GenBank mRNA | JV642456.1 | - | 1022 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
