Assay Details
Gene Symbol: TACSTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114599.4 | XP_001114599.1 | 2312 | 65 | ||
XM_028833244.1 | XP_028689077.1 | 1773 | 65 | |||
GenBank mRNA | JU486679.1 | - | 1117 | 65 | ||
JV651487.1 | - | 729 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
