Assay Details
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842426.1 | XP_028698259.1 | 431 | 75 | ||
XM_028842427.1 | XP_028698260.1 | 1002 | 75 | |||
XM_028842428.1 | XP_028698261.1 | 917 | 75 | |||
GenBank mRNA | JV053177.1 | - | 379 | 75 | ||
JV643674.1 | - | 344 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
