Assay Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115605.2 | XP_014971091.1 | 272 | 93 | ||
| GenBank mRNA | JV489148.1 | - | 126 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLN| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115605.2 | XP_014971091.1 | 272 | 93 | ||
| GenBank mRNA | JV489148.1 | - | 126 | 93 | ||
Set Membership: |
|