Assay Details
| Gene Symbol: ZNF460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801413.3 | XP_002801459.2 | 1621 | 87 | ||
| XM_015125084.2 | XP_014980570.2 | 2142 | 87 | |||
| XM_015125085.2 | XP_014980571.2 | 1969 | 87 | |||
| GenBank mRNA | JU331106.1 | - | 122 | 87 | ||
More Information
Set Membership: |
|

ZNF460
Genomic Map