Assay Details
| Gene Symbol: XIAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128136.2 | XP_014983622.1 | 1444 | 93 | ||
| XM_015128137.2 | XP_014983623.1 | 1344 | 93 | |||
| XM_015128138.2 | XP_014983624.1 | 1418 | 93 | |||
| XM_028842530.1 | XP_028698363.1 | 1161 | 93 | |||
| GenBank mRNA | JU334299.1 | - | 1185 | 93 | ||
| JU475407.1 | - | 1139 | 93 | |||
More Information
Set Membership: |
|

XIAP
Genomic Map