Assay Details
| Gene Symbol: LOC114670386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827963.1 | XP_028683796.1 | 1608 | 80 | ||
| XM_028827964.1 | XP_028683797.1 | 1528 | 80 | |||
| XM_028827965.1 | XP_028683798.1 | 2214 | 80 | |||
| GenBank mRNA | JU489818.1 | - | 224 | 80 | ||
| Gene Symbol: PRODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149690.2 | XP_015005176.2 | 2164 | 80 | ||
| GenBank mRNA | JU325172.1 | - | 2146 | 80 | ||
| JV622442.1 | - | 2152 | 80 | |||
More Information
Set Membership: |
|

LOC114670386
Genomic Map