Assay Details
Gene Symbol: LOC720399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114093.2 | XP_014969579.1 | 145 | 87 | ||
XM_015114097.2 | XP_014969583.1 | 152 | 87 | |||
XM_028826641.1 | XP_028682474.1 | 188 | 87 | |||
XM_028826644.1 | XP_028682477.1 | 152 | 87 | |||
GenBank mRNA | JU334904.1 | - | 89 | 87 | ||
JU475909.1 | - | 118 | 87 | |||
JU475910.1 | - | 118 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015114093.2, XM_015114097.2, XM_028826641.1, XM_028826644.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
