Assay Details
| Gene Symbol: NANOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826912.1 | XP_028682745.1 | 1486 | 75 | ||
| GenBank mRNA | JU342821.1 | - | 524 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NANOS1| Gene Symbol: NANOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826912.1 | XP_028682745.1 | 1486 | 75 | ||
| GenBank mRNA | JU342821.1 | - | 524 | 75 | ||
Set Membership: |
|