Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123209.2 | XP_014978695.1 | 634 | 67 | ||
GenBank mRNA | JU320747.1 | - | 193 | 67 | ||
JU471248.1 | - | 233 | 67 | |||
JV045564.1 | - | 220 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
