Assay Details
| Gene Symbol: ADRA2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137909.2 | XP_014993395.2 | 382 | 112 | ||
| GenBank mRNA | JV572469.1 | - | 126 | 112 | ||
| JV707871.1 | - | 96 | 112 | |||
More Information
Set Membership: |
|

ADRA2C
Genomic Map