Assay Details
Gene Symbol: LOC719082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028826307.1 | XP_028682140.1 | 188 | 96 | ||
XM_028826308.1 | XP_028682141.1 | 188 | 96 | |||
XM_028826309.1 | XP_028682142.1 | 188 | 96 | |||
XM_028826310.1 | XP_028682143.1 | 188 | 96 | |||
XM_028826311.1 | XP_028682144.1 | 188 | 96 | |||
XM_028826312.1 | XP_028682145.1 | 188 | 96 | |||
XM_028826313.1 | XP_028682146.1 | 188 | 96 | |||
XM_028826314.1 | XP_028682147.1 | 188 | 96 | |||
XM_028826315.1 | XP_028682148.1 | 188 | 96 | |||
XM_028826316.1 | XP_028682149.1 | 188 | 96 | |||
GenBank mRNA | JU333782.1 | - | 97 | 96 | ||
JU334443.1 | - | 97 | 96 | |||
JU475538.1 | - | 101 | 96 | |||
JV611795.1 | - | 110 | 96 | |||
JV711218.1 | - | 101 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826307.1, XM_028826308.1, XM_028826309.1, XM_028826310.1, XM_028826311.1, XM_028826312.1, XM_028826313.1, XM_028826314.1, XM_028826315.1, XM_028826316.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
