Assay Details
Gene Symbol: LOC719082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028826307.1 | XP_028682140.1 | 87 | |||
XM_028826308.1 | XP_028682141.1 | 87 | ||||
XM_028826309.1 | XP_028682142.1 | 87 | ||||
XM_028826310.1 | XP_028682143.1 | 87 | ||||
XM_028826311.1 | XP_028682144.1 | 87 | ||||
XM_028826312.1 | XP_028682145.1 | 87 | ||||
XM_028826313.1 | XP_028682146.1 | 87 | ||||
XM_028826314.1 | XP_028682147.1 | 87 | ||||
XM_028826315.1 | XP_028682148.1 | 87 | ||||
XM_028826316.1 | XP_028682149.1 | 87 | ||||
GenBank mRNA | JU333782.1 | - | 87 | |||
JU334443.1 | - | 87 | ||||
JU475538.1 | - | 221 | 87 | |||
JV047017.1 | - | 87 | ||||
JV611795.1 | - | 230 | 87 | |||
JV711218.1 | - | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
