Assay Details
| Gene Symbol: FAM111A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090885.4 | XP_001090885.4 | 2880 | 141 | ||
| XM_015114129.2 | XP_014969615.2 | 3134 | 141 | |||
| XM_015114130.2 | XP_014969616.2 | 3173 | 141 | |||
| XM_015114131.2 | XP_014969617.2 | 2949 | 141 | |||
| XM_028833396.1 | XP_028689229.1 | 2988 | 141 | |||
| XM_028833397.1 | XP_028689230.1 | 2831 | 141 | |||
| GenBank mRNA | JU491319.1 | - | 347 | 141 | ||
| JU491321.1 | - | 417 | 141 | |||
More Information
Set Membership: |
|

FAM111A
Genomic Map