Assay Details
| Gene Symbol: ZNF195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113393.2 | XP_014968879.2 | 730 | 78 | ||
| XM_015113394.2 | XP_014968880.2 | 627 | 78 | |||
| XM_015113395.2 | XP_014968881.2 | 547 | 78 | |||
| XM_028833083.1 | XP_028688916.1 | 731 | 78 | |||
| GenBank mRNA | JU328927.1 | - | 50 | 78 | ||
| JV070745.1 | - | 70 | 78 | |||
More Information
Set Membership: |
|

ZNF195
Genomic Map