Assay Details
Gene Symbol: LOC698860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845698.1 | XP_028701531.1 | 2793 | 123 | ||
XM_028845699.1 | XP_028701532.1 | 2633 | 123 | |||
XM_028845700.1 | XP_028701533.1 | 1450 | 123 | |||
GenBank mRNA | JU334459.1 | - | 1066 | 123 | ||
JU475548.1 | - | 982 | 123 | |||
JV043323.1 | - | 1152 | 123 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
