Assay Details
Gene Symbol: RAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082554.3 | XP_001082554.1 | 1925 | 117 | ||
XM_028841908.1 | XP_028697741.1 | 1832 | 117 | |||
GenBank mRNA | JV050039.1 | - | 521 | 117 | ||
JV640578.1 | - | 1506 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
