Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808430.3 | XP_002808476.3 | 1926 | 104 | ||
| GenBank mRNA | JV715036.1 | - | 815 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD180| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808430.3 | XP_002808476.3 | 1926 | 104 | ||
| GenBank mRNA | JV715036.1 | - | 815 | 104 | ||
Set Membership: |
|