Assay Details
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142607.2 | XP_014998093.2 | 5165 | 112 | ||
XM_015142613.2 | XP_014998099.2 | 5102 | 112 | |||
XM_015142624.2 | XP_014998110.2 | 4851 | 112 | |||
GenBank mRNA | JU324991.1 | - | 1532 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
