Assay Details
| Gene Symbol: PLVAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123258.2 | XP_014978744.2 | 2410 | 87 | ||
| GenBank mRNA | JU494480.1 | - | 220 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLVAP| Gene Symbol: PLVAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123258.2 | XP_014978744.2 | 2410 | 87 | ||
| GenBank mRNA | JU494480.1 | - | 220 | 87 | ||
Set Membership: |
|