Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139993.2 | XP_014995479.1 | 2003 | 88 | ||
| GenBank mRNA | JU328357.1 | - | 220 | 88 | ||
| JV636320.1 | - | 651 | 88 | |||
More Information
Set Membership: |
|

TMEM267
Genomic Map