Assay Details
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002799323.3 | XP_002799369.2 | 5671 | 112 | ||
GenBank mRNA | JU337856.1 | - | 825 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
