Assay Details
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135311.2 | XP_014990797.1 | 1912 | 80 | ||
XM_028846488.1 | XP_028702321.1 | 1912 | 80 | |||
GenBank mRNA | JV580665.1 | - | 141 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
