Assay Details
| Gene Symbol: SOX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118518.2 | XP_014974004.2 | 1274 | 83 | ||
| GenBank mRNA | JV659569.1 | - | 323 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX15| Gene Symbol: SOX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118518.2 | XP_014974004.2 | 1274 | 83 | ||
| GenBank mRNA | JV659569.1 | - | 323 | 83 | ||
Set Membership: |
|