Assay Details
Gene Symbol: PPP1R9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119321.2 | XP_014974807.1 | 277 | 86 | ||
GenBank mRNA | JU324890.1 | - | 127 | 86 | ||
JV631215.1 | - | 106 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
