Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105001.4 | XP_001105001.2 | 1198 | 107 | ||
GenBank mRNA | JU325882.1 | - | 923 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
