Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149166.2 | XP_015004652.1 | 84 | |||
| XM_028828340.1 | XP_028684173.1 | 84 | ||||
| GenBank mRNA | JU326340.1 | - | 47 | 84 | ||
| JV045376.1 | - | 84 | ||||
| JV490425.1 | - | 84 | ||||
| JV602237.1 | - | 84 | ||||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028828340.1 transcripts.
Set Membership: |
|

DYNLRB1
Genomic Map