Assay Details
Gene Symbol: LOC719678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842716.1 | XP_028698549.1 | 408 | 60 | ||
GenBank mRNA | JV532383.1 | - | 168 | 60 |
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828710.1 | XP_028684543.1 | 291 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
