Assay Details
Gene Symbol: PRR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122709.2 | XP_014978195.1 | 4297 | 80 | ||
XM_028839973.1 | XP_028695806.1 | 4229 | 80 | |||
GenBank mRNA | JV662082.1 | - | 121 | 80 | ||
JV697116.1 | - | 1499 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
