Assay Details
Gene Symbol: C14H11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028833319.1 | XP_028689152.1 | 164 | 66 | ||
XM_028833320.1 | XP_028689153.1 | 164 | 66 | |||
XM_028833321.1 | XP_028689154.1 | 163 | 66 | |||
GenBank mRNA | JV626570.1 | - | 53 | 66 | ||
JV694088.1 | - | 64 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
