Assay Details
| Gene Symbol: SAP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134472.2 | XP_014989958.2 | 413 | 95 | ||
| XM_028845597.1 | XP_028701430.1 | 379 | 95 | |||
| GenBank mRNA | JV045824.1 | - | 97 | 95 | ||
| JV639603.1 | - | 93 | 95 | |||
More Information
Set Membership: |
|

SAP25
Genomic Map