Assay Details
Gene Symbol: BEND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847303.1 | XP_028703136.1 | 100 | 87 | ||
GenBank mRNA | JV067186.1 | - | 54 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028847303.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
