Assay Details
| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU336981.1 | - | 83 | 68 | ||
| JV726283.1 | - | 72 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOL3| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU336981.1 | - | 83 | 68 | ||
| JV726283.1 | - | 72 | 68 | |||
Set Membership: |
|