Assay Details
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082027.4 | XP_001082027.4 | 909 | 88 | ||
XM_028851267.1 | XP_028707100.1 | 913 | 88 | |||
GenBank mRNA | JV641971.1 | - | 271 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
