Assay Details
Gene Symbol: MEPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099140.4 | XP_001099140.1 | 183 | 98 | ||
XM_028848610.1 | XP_028704443.1 | 184 | 98 | |||
GenBank mRNA | JV513606.1 | - | 93 | 98 | ||
JV719163.1 | - | 102 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
