Assay Details
| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141220.2 | XP_014996706.2 | 3471 | 111 | ||
| GenBank mRNA | JV648447.1 | - | 226 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL17B| Gene Symbol: IL17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141220.2 | XP_014996706.2 | 3471 | 111 | ||
| GenBank mRNA | JV648447.1 | - | 226 | 111 | ||
Set Membership: |
|