Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150033.2 | XP_015005519.2 | 1130 | 73 | ||
| XM_028827868.1 | XP_028683701.1 | 1126 | 73 | |||
| XM_028827869.1 | XP_028683702.1 | 1126 | 73 | |||
| XM_028827870.1 | XP_028683703.1 | 1126 | 73 | |||
| XM_028827871.1 | XP_028683704.1 | 1126 | 73 | |||
| GenBank mRNA | JV617644.1 | - | 1146 | 73 | ||
More Information
Set Membership: |
|

NEFH
Genomic Map