Assay Details
Gene Symbol: RNASE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090353.4 | XP_001090353.1 | 1188 | 104 | ||
XM_015143210.2 | XP_014998696.1 | 658 | 104 | |||
GenBank mRNA | JV059727.1 | - | 314 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
