Assay Details
Gene Symbol: LOC699231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126840.2 | XP_014982326.2 | 2314 | 82 | ||
GenBank mRNA | JV670454.1 | - | 236 | 82 | ||
JV705170.1 | - | 139 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
