Assay Details
Gene Symbol: LOC114669857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU350285.1 | - | 119 | 88 |
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131300.2 | XP_014986786.2 | 126 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131300.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
