Assay Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852029.1 | XP_028707862.1 | 897 | 94 | ||
| XM_028852030.1 | XP_028707863.1 | 841 | 94 | |||
| GenBank mRNA | JV467714.1 | - | 222 | 94 | ||
| JV714306.1 | - | 208 | 94 | |||
More Information
Set Membership: |
|

HDDC3
Genomic Map